missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10732695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10732695 25 g Glass bottle 25g
10379200 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732695 Supplier Thermo Scientific Acros Supplier No. 294140250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 5527-95-7 | Molecular Formula: C7H4ClFO | Molecular Weight: 158.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5527-95-7
MDL Number MFCD00143288
InChI Key AZMDWRPTDCIFRD-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorobenzaldehyde, 4-chloro-3-fluorobenz-aldehyde, 4-chloro-3-fluoro-benzaldehyde, benzaldehyde, 4-chloro-3-fluoro, pubchem1412, acmc-1amt7, ksc494a8n, 4-chloro-3-fluoro benzaldehyde, azmdwrptdcifrd-uhfffaoysa
PubChem CID 2724985
IUPAC Name 4-chloro-3-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)Cl

Spécification

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 49°C
Color White to Yellow
Flash Point 96.7°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H4ClFO
Linear Formula ClC6H3(F)CHO
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Molecular Weight (g/mol) 158.56
Formula Weight 158.56
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 4-Chloro-3-fluorobenzaldehyde
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.