missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10379200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10379200 5 g Glass bottle 5g
10732695 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10379200 Supplier Thermo Scientific Acros Supplier No. 294140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5527-95-7
MDL Number MFCD00143288
InChI Key AZMDWRPTDCIFRD-UHFFFAOYSA-N
Synonym 3-fluoro-4-chlorobenzaldehyde, 4-chloro-3-fluorobenz-aldehyde, 4-chloro-3-fluoro-benzaldehyde, benzaldehyde, 4-chloro-3-fluoro, pubchem1412, acmc-1amt7, ksc494a8n, 4-chloro-3-fluoro benzaldehyde, azmdwrptdcifrd-uhfffaoysa
PubChem CID 2724985
IUPAC Name 4-chloro-3-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)F)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4ClFO
Linear Formula ClC6H3(F)CHO
Quantity 5 g
Molecular Weight (g/mol) 158.556
Formula Weight 158.56
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-3-fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.