missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-fluoroaniline, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-3-fluoroaniline |
| CAS | 367-22-6 |
| Molecular Formula | C6 H5 Cl F N |
| Quantity | 1 g |
| Synonym | 4-Chloro-3-fluoro-aniline, 3-Fluoro-4-chloroaniline, 3-Fluoro-4-chlorobenzenamine, 4-Chloro-3-fluorobenzenamine, 4-Chloro-3-fluoro-benzeneamine, 4-Chloro-3-fluorophenylamine |
| InChI Formula | InChI=1S/C6H5ClFN/c7-5-2-1-4(9)3-6(5)8/h1-3H,9H2 |
| SMILES | Nc1ccc(Cl)c(F)c1 |
| IUPAC Name | 4-chloro-3-fluoroaniline |
| Molecular Weight (g/mol) | 145.56 |
| Formula Weight | 145.0095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction