missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 15498765
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1323955-63-0
Molecular Formula C10H10ClFO3
Molecular Weight (g/mol) 232.635
MDL Number MFCD19687188
InChI Key JLMJTUGWXPDCKF-UHFFFAOYSA-N
Synonym 4-chloro-3-ethoxy-2-fluorophenylacetic acid, 4-chloro-3-ethoxy-2-fluorophenyl acetic acid, 2-4-chloro-3-ethoxy-2-fluorophenyl acetic acid
PubChem CID 66523623
IUPAC Name 2-(4-chloro-3-ethoxy-2-fluorophenyl)acetic acid
SMILES CCOC1=C(C=CC(=C1F)CC(=O)O)Cl

Specifications

Quantity 250 mg
Physical Form Solid
Formula Weight 232.64
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-ethoxy-2-fluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'