missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-chloro-3-cyanophenylboronic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 300.0°C to 304.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Formula Weight | 181.39 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 4-Chloro-3-cyanophenylboronic acid
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'