missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3,5-dinitrobenzonitrile, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-3,5-dinitrobenzonitrile |
| CAS | 1930-72-9 |
| Molecular Formula | C7H2ClN3O4 |
| Quantity | 100 mg |
| Synonym | 3,5-Dinitro-4-chlorobenzonitrile, 4-Chloro-3,5-dinitrobenzonitrile, NSC 74453 |
| InChI Formula | InChI=1S/C7H2ClN3O4/c8-7-5(10(12)13)1-4(3-9)2-6(7)11(14)15/h1-2H |
| SMILES | N#CC1=CC([N+]([O-])=O)=C(Cl)C([N+]([O-])=O)=C1 |
| IUPAC Name | 4-chloro-3,5-dinitrobenzonitrile |
| Molecular Weight (g/mol) | 227.56 |
| Formula Weight | 226.97 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction