missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzoic acid, 97%

Product Code. 11479513
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479513 100 g 100g
11469513 25 g 25g
11489513 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479513 Supplier Thermo Scientific Alfa Aesar Supplier No. A12373.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 118-97-8 | C7H3ClN2O6 | 246.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-97-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.559
MDL Number MFCD00007080
InChI Key PCTFIHOVQYYAMH-UHFFFAOYSA-N
Synonym 4-chloro-3,5-dinitrobenzoic acid, 3,5-dinitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzoate, 4-chloro-3,5-dinitro-benzoic acid, benzoicacid, 4-chloro-3,5-dinitro, pubchem22980, acmc-1bt9j, ksc180k5t, timtec-bb sbb003176
PubChem CID 8377
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 159°C to 162°C
Boiling Point 315°C
Flash Point 186°C (367°F)
Quantity 100 g
Beilstein 668253
Formula Weight 246.56
Percent Purity 99%
Chemical Name or Material 4-Chloro-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.