missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)pyridine, 98%

Product Code. 15492209
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492209 1 g 1g
15412219 250 mg 250mg
15402219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492209 Supplier Thermo Scientific Alfa Aesar Supplier No. H64833.03
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131748-14-6 | C6H3ClF3N | 181.54 g/mol

Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131748-14-6
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD07368047
InChI Key FZINIBTZNSPWQR-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine
PubChem CID 14761442
IUPAC Name 4-chloro-2-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=NC=CC(Cl)=C1

Specifications

Color Colorless
Quantity 1 g
UN Number UN2810
Solubility Information Slightly soluble in water.
Formula Weight 181.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.