missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)phenylboronic acid, 95%, Thermo Scientific™

Product Code. 11996131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11996131 5 g 5g
11976131 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996131 Supplier Thermo Scientific Acros Supplier No. 445380050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 313545-41-4
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.38
InChI Key YAPBOBGBYQQYHX-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethylphenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl-phenylboronic acid, boronic acid, 4-chloro-2-trifluoromethyl phenyl, 4-chloro-2-trifluoromethyl phenylboronicacid, pubchem5158, acmc-20aic7
PubChem CID 2763244
IUPAC Name [4-chloro-2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O

Specifications

Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 224.38
Percent Purity 95%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)phenylboronic acid
compliance-icons
Product Identifier
  • 4-Chloro-2-(trifluoromethyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.