missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethoxy)phenylacetonitrile, 97%, Thermo Scientific™

Catalog Number 15448635
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1261861-63-5
C9H5ClF3NO
235.59
MFCD18393878
VOLGTZZPDCMHPW-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy phenylacetonitrile, 2-4-chloro-2-trifluoromethoxy phenyl acetonitrile
66523574
2-[4-chloro-2-(trifluoromethoxy)phenyl]acetonitrile
C1=CC(=C(C=C1Cl)OC(F)(F)F)CC#N
This item is not returnable. View return policy
1261861-63-5
C9H5ClF3NO
235.59
MFCD18393878
VOLGTZZPDCMHPW-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy phenylacetonitrile, 2-4-chloro-2-trifluoromethoxy phenyl acetonitrile
66523574
2-[4-chloro-2-(trifluoromethoxy)phenyl]acetonitrile
C1=CC(=C(C=C1Cl)OC(F)(F)F)CC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.472
Quantity 250 mg
UN Number UN3276
Formula Weight 235.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'