missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10367444
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367444 5 g Amber Glass Bottle 5g
10521322 250 mg Amber Glass Bottle 250mg
10622073 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367444 Supplier Thermo Scientific Maybridge Supplier No. SEW03561DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-77-3
Molecular Formula C7H5ClF3NO
Molecular Weight (g/mol) 211.568
MDL Number MFCD00153102
InChI Key UCFTYLMKCJPWBX-UHFFFAOYSA-N
PubChem CID 2781526
IUPAC Name 4-chloro-2-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1Cl)OC(F)(F)F)N

Specifications

CAS Min % 97.0
Quantity 5 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Synonym 4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine
Formula Weight 211.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.