missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-2-(trifluoromethoxy)aniline is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water 0.25 g/L @25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
4-Chloro-2-(trifluoromethoxy)aniline is used as a pharmaceutical intermediate.
Solubility
Sparingly soluble in water 0.25 g/L @25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 31°C to 33°C |
Color | White |
UN Number | UN2811 |
Synonym | 4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine |
Solubility Information | Sparingly soluble in water 0.25g/L @25°C. |
Formula Weight | 211.57 |
Percent Purity | 97% |
Boiling Point | 82°C (15 mmHg) |
Quantity | 1 g |
Physical Form | Crystalline Powder |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'