missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrophenol, 98%, contains up to ca 10% water, Thermo Scientific Chemicals

Product Code. 11413667
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11413667

Brand: Thermo Scientific Alfa Aesar A18150.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-64-5
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00007113
InChI Key NWSIFTLPLKCTSX-UHFFFAOYSA-N
Synonym 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol
PubChem CID 6980
IUPAC Name 4-chloro-2-nitrophenol
SMILES OC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 85°C to 88°C
Quantity 25 g
Beilstein 2048031
Formula Weight 173.56
Percent Purity 98%
Assay contains up to ≈10% water
Chemical Name or Material 4-Chloro-2-nitrophenol
compliance-icons
Product Identifier
  • 4-Chloro-2-nitrophenol, 98%, contains up to ca 10% water
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-Chloro-2-nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.