missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrobenzyl chloride, 98%

Product Code. 10001764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10001764 1 g 1g
10396314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001764 Supplier Thermo Scientific Acros Supplier No. 187950010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 938-71-6 | C7H5Cl2NO2 | 206.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-71-6
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.02
MDL Number MFCD00007216
InChI Key QBIQAXWMONJLNW-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrobenzyl chloride, 4-chloro-1-chloromethyl-2-nitrobenzene, 4-chloro-2-nitrobenzylchloride, acmc-20amlj, benzene, 4-chloro-1-chloromethyl-2-nitro, 2-nitro-4-chlorobenzyl chloride, omphlgrocarzou-uhfffaoysa, .alpha.,4-dichloro-2-nitrotoluene
PubChem CID 598399
SMILES [O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1

Specifications

Melting Point 38°C to 44°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H3(NO2)CH2Cl
Quantity 1 g
IUPAC Name 4-chloro-1-(chloromethyl)-2-nitrobenzene
Formula Weight 206.03
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-nitrobenzyl chloride
compliance-icons
Product Identifier
  • 4-Chloro-2-nitrobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.