missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrobenzoic acid, 97%

Product Code. 11485303
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485303 100 g 100g
11475303 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485303 Supplier Thermo Scientific Alfa Aesar Supplier No. A12872.22
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6280-88-2 | C7H4ClNO4 | 201.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6280-88-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.56
MDL Number MFCD00007214
InChI Key JAHIPDTWWVYVRV-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrobenzoic acid, 2-nitro-4-chlorobenzoic acid, 4-chloro-2-nitrobenzoicacid, benzoic acid, 4-chloro-2-nitro, 4-chloro-2-nitro-benzoic acid, acmc-1b6vn, ksc353a2d, 2-carboxy-5-chloronitrobenzene, benzoicacid, 4-chloro-2-nitro, 2-nitro-4-chlorobenzoic acid anion
PubChem CID 80475
IUPAC Name 4-chloro-2-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 142°C to 146°C
Flash Point >100°C (212°F)
Odor Odorless
Quantity 100 g
Beilstein 2051370
Formula Weight 201.57
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.