missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitroaniline, TRC

Product Code. 30362679
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30362679 1 g -
30356418 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 30362679

Brand: Toronto Research Chemicals TRCC3695551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-63-4
Molecular Formula C6 H5 Cl N2 O2
Molecular Weight (g/mol) 172.57
Synonym Aniline, 4-chloro-2-nitro- (8CI), 4-Chloro-2-nitrobenzenamine, (4-Chloro-2-nitrophenyl)amine, 1-Amino-2-nitro-4-chlorobenzene, 2-Nitro-4-chloroaniline, 4-Chloro-2-nitroaniline, Azoene Fast Red 3GL Base, Daito Red Base 3GL, Devol Red F, Diazo Fast Red 3GL, Fast Red 2NC Base, Fast Red 3GL Base, Fast Red 3GL Special Base, Fast Red Base 3JL, Hiltonil Fast Red 3GL Base, Kayaku Fast Red 3GL Base, Mitsui Red 3GL Base, NSC 3546, Naphthanil Red 3G Base, Naphtoelan Fast Red 3GL Base, PCON, PCONA, Red 3G base, Red Base 3GL, Red Base Ciba VI, Red Base Irga VI, Shinnippon Fast Red 3GL Base, p-Chloro-o-nitroaniline
IUPAC Name 4-chloro-2-nitroaniline
SMILES Nc1ccc(Cl)cc1[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H5ClN2O2/c7-4-1-2-5(8)6(3-4)9(10)11/h1-3H,8H2
Formula Weight 172.004
Chemical Name or Material 4-Chloro-2-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.