missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitroaniline, 98%

Product Code. 11463657
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463657 5000 g 5000g
11453657 250 g 250g
11443657 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463657 Supplier Thermo Scientific Alfa Aesar Supplier No. A18143.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 89-63-4 | C6H5ClN2O2 | 172.57 g/mol

4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

Solubility
Soluble in water (0.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-63-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00007836
InChI Key PBGKNXWGYQPUJK-UHFFFAOYSA-N
Synonym benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi
PubChem CID 6979
IUPAC Name 4-chloro-2-nitroaniline
SMILES NC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 114°C to 118°C
Density 1.37
Flash Point 191°C (375°F)
Odor Weak, Characteristic
Quantity 5000 g
UN Number UN2237
Beilstein 512436
Solubility Information Soluble in water (0.5g/L).
Formula Weight 172.57
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-nitroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.