missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(methylthio)pyrimidine, 98%

Product Code. 11497014
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497014 250 g 250g
11477014 10 g 10g
11487014 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497014 Supplier Thermo Scientific Alfa Aesar Supplier No. B21481.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 49844-90-8 | C5H5ClN2S | 160.62 g/mol

It finds it application as a building block used in medicinal chemistry synthesis. 4-Chloro-2-methylthiopyrimidine is used in the total synthesis of the marine alkaloid variolin B1 and 2-hydroxy-4-pyrimidinecarboxylic acid. It is also used in the synthesis of 2,4-disubstituted pyrimidines, a novel class of KDR kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a building block used in medicinal chemistry synthesis. 4-Chloro-2-methylthiopyrimidine is used in the total synthesis of the marine alkaloid variolin B1 and 2-hydroxy-4-pyrimidinecarboxylic acid. It is also used in the synthesis of 2,4-disubstituted pyrimidines, a novel class of KDR kinase inhibitors.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture, heat and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49844-90-8
Molecular Formula C5H5ClN2S
Molecular Weight (g/mol) 160.62
MDL Number MFCD00006083
InChI Key DFOHHQRGDOQMKG-UHFFFAOYSA-N
Synonym 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
PubChem CID 97113
IUPAC Name 4-chloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC=CC(Cl)=N1

Specifications

Melting Point ∼-2°C
Density 1.38
Boiling Point 118°C to 120°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6 00
Quantity 250 g
UN Number UN1760
Beilstein 118042
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 160.63
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-(methylthio)pyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.