missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylphenyl isocyanate, 98%

Product Code. 11378445
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11378445

Brand: Thermo Scientific Alfa Aesar L10470.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37408-18-7 | C8H6ClNO | 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37408-18-7
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00019914
InChI Key FTZJLXIATZSKIL-UHFFFAOYSA-N
Synonym 4-chloro-2-methylphenyl isocyanate, 4-chloro-2-methylphenylisocyanate, 4-chloro-o-tolyl isocyanate, benzene, 4-chloro-1-isocyanato-2-methyl, 4-chloro-1-isocyanato-2-methyl-benzene, acmc-20amlp, 4-chloro-2-methylbenzenisocyanate, #, 4-chloranyl-1-isocyanato-2-methyl-benzene
PubChem CID 142157
IUPAC Name 4-chloro-1-isocyanato-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)N=C=O

Specifications

Melting Point 36°C to 40°C
Boiling Point 103°C (13 mmHg)
Quantity 5 g
UN Number UN2811
Beilstein 2803097
Formula Weight 167.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylphenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.