missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methyl-6-nitroaniline, 97%, Thermo Scientific™

Product Code. 10534721 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10534721 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10534721 Supplier Thermo Scientific Supplier No. 326400050
 Restricted Item
Estimated Shipment: 26-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62790-50-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 126°C to 132°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7ClN2O2
MDL Number MFCD01320687
Quantity 5 g
Synonym 4-chloro-2-methyl-6-nitrophenylamine, acmc-1b9ic, 2-amino-5-chloro-3-nitrotoluene, 2-methyl-4-chloro-6-nitroaniline, 4-chloro-2-methyl-6-nitrobenzenamine, 2-amino-5-chloro-3-methylnitrobenzene, benzenamine,4-chloro-2-methyl-6-nitro, benzenamine, 4-chloro-2-methyl-6-nitro
InChI Key QDSCDFKGUAONPC-UHFFFAOYSA-N
SMILES CC1=CC(Cl)=CC(=C1N)[N+]([O-])=O
IUPAC Name 4-chloro-2-methyl-6-nitroaniline
Molecular Weight (g/mol) 186.60
PubChem CID 182355
Formula Weight 186.6
Percent Purity 97%
Physical Form Crystals or Needles
Chemical Name or Material 4-Chloro-2-methyl-6-nitroaniline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-2-methyl-6-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.