missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxyphenol, 99%, Thermo Scientific™

Product Code. 10081751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081751 10 g Glass bottle 10g
10099932 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081751 Supplier Thermo Scientific Acros Supplier No. 294110100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16766-30-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.58
MDL Number MFCD00070774
InChI Key FVZQMMMRFNURSH-UHFFFAOYSA-N
PubChem CID 28050
IUPAC Name 4-chloro-2-methoxyphenol
SMILES COC1=CC(Cl)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16.0°C to 17.0°C
Color Yellow to Brown
Density 1.3000g/mL
Boiling Point 241.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H3(OCH3)OH
Refractive Index 1.5575 to 1.5595
Quantity 10 g
Specific Gravity 1.3
Synonym 4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa
Formula Weight 158.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methoxyphenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-2-methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.