missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 10654165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10654165 5 g 5g
10326344 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10654165 Supplier Thermo Scientific Acros Supplier No. 398560050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90296-27-8
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.61
MDL Number MFCD02683547
InChI Key AXXPZAITCCIOIA-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
PubChem CID 3549393
IUPAC Name (4-chloro-2-methoxyphenyl)methanol
SMILES COC1=C(CO)C=CC(Cl)=C1

Specifications

Melting Point 47°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula ClC6H3(OH)OCH3
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxybenzyl alcohol
compliance-icons
Product Identifier
  • 4-Chloro-2-methoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.