missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-isopropyl-5-methylphenol, TRC

Product Code. 30350569
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350569 500 mg 500mg
30359774 5 g 5g
30364692 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350569 Supplier Toronto Research Chemicals Supplier No. TRCC597088500MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-68-9
Molecular Formula C10 H13 Cl O
Molecular Weight (g/mol) 184.66
Synonym Thymol, 6-chloro- (6CI,7CI,8CI), 4-Chloro-5-methyl-2-(1-methylethyl)phenol, 2-Isopropyl-4-chloro-5-methylphenol, 4-Chloro-2-isopropyl-5-methylphenol, 4-Chloro-5-methyl-2-(propan-2-yl)phenol, 4-Chloro-6-isopropyl-3-methylphenol, 6-Chlorothymol, Chlorothymol, Chlorthymol, NSC 406261, NSC 4964
IUPAC Name 4-chloro-5-methyl-2-propan-2-ylphenol
SMILES CC(C)c1cc(Cl)c(C)cc1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H13ClO/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6,12H,1-3H3
Formula Weight 184.0655
Chemical Name or Material Chlorothymol (4-Chloro-2-isopropyl-6-methylphenol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.