missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodoaniline, 98+%, Thermo Scientific Chemicals

Product Code. 10153194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153194 25 g Glass Bottle 25g
10411284 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153194 Supplier Thermo Scientific Acros Supplier No. 396840250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63069-48-7
MDL Number MFCD01863737
InChI Key FLEJOBRWKBPUOX-UHFFFAOYSA-N
Synonym 2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
PubChem CID 2757615
IUPAC Name 4-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)I)N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C6H5ClIN
Linear Formula ClC6H3(I)NH2
Quantity 25 g
Molecular Weight (g/mol) 253.467
Formula Weight 253.47
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 4-chloro-2-iodoaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-2-iodoaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.