missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15447316
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765917-27-9
Molecular Formula C12H15BClFO2
Molecular Weight (g/mol) 256.508
InChI Key HWHYSIMGKXYKDS-UHFFFAOYSA-N
Synonym 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl, 4-chloro-2-fluorophenylboronic aicd, pinacol ester, 2-4-chloro-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 22273588
IUPAC Name 2-(4-chloro-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)Cl)F

Specifications

Quantity 250 mg
Formula Weight 256.51
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'