missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dimethylaniline, 96%

Product Code. 11439477
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11439477

Brand: Thermo Scientific Alfa Aesar B21996.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24596-18-7 | C8H10ClN | 155.625 g/mol

4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-chloro-2,6-dimethylaniline was chosen as a substrate of medium in palladium-catalyzed cyanations of aryl chlorides.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24596-18-7
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD01076576
InChI Key JCLZLZKUISPXDC-UHFFFAOYSA-N
Synonym 4-chloro-2,6-dimethyl-phenylamine, 4-chloro-2,6-dimethylbenzenamine, 2,6-dimethyl-4-chloroaniline, intermediates-zcf02661, acmc-20a53k, 4-chlor-2,6-dimethylanilin, pharmabridge p-1739, 4-chloro-2,6-dimethyl-aniline, 4-chloro-2,6-xylidine, 4-chloro-2,6-dimethylphenylamine
PubChem CID 5073362
IUPAC Name 4-chloro-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)Cl

Specifications

Melting Point 45°C to 50°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 155.63
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,6-dimethylaniline
compliance-icons
Product Identifier
  • 4-Chloro-2,6-dimethylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Chloro-2,6-dimethylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.