missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-dimethyl-3-nitropyridine, 98%, Thermo Scientific™

Product Code. 15411279
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15513-48-1
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.595
MDL Number MFCD08543468
InChI Key PSBGFLWVQDGETK-UHFFFAOYSA-N
Synonym pyridine, 4-chloro-2,6-dimethyl-3-nitro, 4-chloro-3-nitro-2,6-lutidine, pubchem11177, 4-chloranyl-2,6-dimethyl-3-nitro-pyridine
PubChem CID 10702725
IUPAC Name 4-chloro-2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C(=C1)Cl)[N+](=O)[O-])C

Specifications

Melting Point 70°C to 73°C
Color White
Quantity 250 mg
Beilstein 1569403
Formula Weight 186.6
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-2,6-dimethyl-3-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'