missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,6-difluorobenzylamine, 97%, Thermo Scientific™

Product Code. 11411430
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11411430

Brand: Thermo Scientific Alfa Aesar H26446.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518357-43-2
Molecular Formula C7H6ClF2N
Molecular Weight (g/mol) 177.579
MDL Number MFCD06660350
InChI Key YBILGZZEXORCAH-UHFFFAOYSA-N
Synonym 4-chloro-2,6-difluorobenzylamine, 4-chloro-2,6-difluorophenyl methanamine, benzenemethanamine, 4-chloro-2,6-difluoro, 2,6-difluoro-4-chlorobenzylamine, 4-chloro-2,6-difluorophenyl methylamine, 4-chloro-2,6-difluoro-phenyl methanamine, 1-4-chloro-2,6-difluorophenyl methanamine
PubChem CID 17750831
IUPAC Name (4-chloro-2,6-difluorophenyl)methanamine
SMILES C1=C(C=C(C(=C1F)CN)F)Cl

Specifications

Refractive Index 1.5145
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 177.58
Percent Purity 97%
Chemical Name or Material 4-Chloro-2,6-difluorobenzylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.