missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2,6-difluorobenzylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifica
Specifica
| Refractive Index | 1.5145 |
| Quantity | 1 g |
| UN Number | UN2735 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 177.58 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Chloro-2,6-difluorobenzylamine |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto