missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-2',6'-difluoroacetophenone, 97%, Thermo Scientific™

Product Code. 15435467
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15435467

Brand: Thermo Scientific Alfa Aesar H32711.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017777-45-5
Molecular Formula C8H5ClF2O
Molecular Weight (g/mol) 190.574
MDL Number MFCD09832259
InChI Key VEZAUYYYYPJSKR-UHFFFAOYSA-N
Synonym 4'-chloro-2',6'-difluoroacetophenone, 1-4-chloro-2,6-difluorophenyl ethanone, 1-4-chloro-2,6-difluorophenyl ethan-1-one, 1-acetyl-4-chloro-2,6-difluorobenzene
PubChem CID 46737501
IUPAC Name 1-(4-chloro-2,6-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1F)Cl)F

Specifications

Refractive Index 1.5055
Quantity 5 g
Formula Weight 190.58
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2',6'-difluoroacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.