missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-2,5-dimethoxyaniline, 97%, Thermo Scientific™

Product Code. 11935311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11935311 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11935311 Supplier Thermo Scientific Acros Supplier No. 437861000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6358-64-1
Melting Point 118°C to 120°C
Color White to Brown
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H10ClNO2
MDL Number MFCD00014893
Quantity 100 g
Synonym 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718
InChI Key YGUFQYGSBVXPMC-UHFFFAOYSA-N
SMILES COC1=CC(Cl)=C(OC)C=C1N
IUPAC Name 4-chloro-2,5-dimethoxyaniline
Molecular Weight (g/mol) 187.62
PubChem CID 22833
Formula Weight 187.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-chloro-2, 5-dimethoxyaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-2,5-dimethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.