missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(4-morpholinyl)quinazoline, 97%, Thermo Scientific Chemicals

Product Code. 15420169
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39216-67-6
Molecular Formula C12H12ClN3O
Molecular Weight (g/mol) 249.70
MDL Number MFCD07758764
InChI Key UWAUCVLFWGAOGM-UHFFFAOYSA-N
Synonym 4-chloro-2-morpholinoquinazoline, 4-chloro-2-morpholin-4-yl quinazoline, 4-4-chloroquinazolin-2-yl morpholine, 4-chloro-2-4-morpholinyl quinazoline
PubChem CID 12286589
IUPAC Name 4-(4-chloroquinazolin-2-yl)morpholine
SMILES ClC1=C2C=CC=CC2=NC(=N1)N1CCOCC1

Specifications

Quantity 250 mg
Formula Weight 249.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-2-(4-morpholinyl)quinazoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'