missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(4-chlorobenzoyl)-1-(4-methoxyphenyl)hydrazide Benzoic Acid, TRC

Product Code. 30135963
Change view
Click to view available options
Quantity:
50 mg
100 mg
2.5 g
Unit Size:
100mg
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135963 2.5 g 2.5g
30132560 50 mg 50mg
30008623 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135963 Supplier Toronto Research Chemicals Supplier No. TRCC3655402.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402849-27-8
Molecular Formula C21H16Cl2N2O3
Molecular Weight (g/mol) 415.27
Synonym Benzoic acid, 4-chloro-, 2-(4-chlorobenzoyl)-1-(4-methoxyphenyl)hydrazide, 4-Chloro-N'-(4-chlorobenzoyl)-N-(4-methoxyphenyl)benzohydrazide, Indometacin dibenzylate
IUPAC Name 4-chloro-N'-(4-chlorobenzoyl)-N'-(4-methoxyphenyl)benzohydrazide
SMILES COc1ccc(cc1)N(NC(=O)c2ccc(Cl)cc2)C(=O)c3ccc(Cl)cc3

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C21H16Cl2N2O3/c1-28-19-12-10-18(11-13-19)25(21(27)15-4-8-17(23)9-5-15)24-20(26)14-2-6-16(22)7-3-14/h2-13H,1H3,(H,24,26)
Formula Weight 414.0538
Percent Purity >95%
Chemical Name or Material 4-Chloro-2-(4-chlorobenzoyl)-1-(4-methoxyphenyl)hydrazide Benzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.