missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-10-[(dimethylamino)propyl]phenothiazine Hydrochloride, TRC

Product Code. 30362688
Change view
Click to view available options
Quantity:
1 mg
25 mg
50 mg
Unit Size:
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362688 25 mg 25mg
30356984 50 mg 50mg
30360504 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362688 Supplier Toronto Research Chemicals Supplier No. TRCC42886025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72372-62-4
Molecular Formula C17H19ClN2S . HCl
Molecular Weight (g/mol) 355.3
Synonym Chlorpromazine Impurity F, 4-Chloro-N,N-dimethyl-10H-phenothiazine-10-propanamine Monohydrochloride, 4-Chloro-N,N-dimethyl-10H-phenothiazine-10-propanamine Hydrochloride
IUPAC Name 3-(4-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3c(Cl)cccc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19ClN2S.ClH/c1-19(2)11-6-12-20-14-8-3-4-10-16(14)21-17-13(18)7-5-9-15(17)20;/h3-5,7-10H,6,11-12H2,1-2H3;1H
Formula Weight 354.0724
Chemical Name or Material 4-Chloro-10-[(dimethylamino)propyl]phenothiazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.