missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-napthalenecarboxylic Acid, TRC

Product Code. 30357738
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357738 1 g 1g
30352021 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357738 Supplier Toronto Research Chemicals Supplier No. TRCC3709401G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-1-napthalenecarboxylic Acid
CAS 1013-04-3
Molecular Formula C11 H7 Cl O2
Quantity 1 g
Synonym 4-Chloro-1-naphthalenecarboxylic Acid, 4-Chloro-1-naphthoic Acid
InChI Formula InChI=1S/C11H7ClO2/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H,(H,13,14)
SMILES OC(=O)c1ccc(Cl)c2ccccc12
IUPAC Name 4-chloronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 206.63
Formula Weight 206.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.