missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-naphthol, 97%

Product Code. 15750789 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15750789

Brand: Thermo Scientific Acros 109551000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 604-44-4 | C10H7ClO | 178.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-44-4
Molecular Formula C10H7ClO
Molecular Weight (g/mol) 178.62
MDL Number MFCD00003974
InChI Key LVSPDZAGCBEQAV-UHFFFAOYSA-N
Synonym 4-chloro-1-naphthol, 1-chloro-4-hydroxynaphthalene, 1-naphthalenol, 4-chloro, 4-chloro-1-naphthalenol, 1-naphthol, 4-chloro, 4-chloronaphthol, 4-chloro-.alpha.-naphthol, 4-chloro-1-hydroxynaphthalene, 4-chloro-alpha-naphthol, acmc-20amm7
PubChem CID 11787
IUPAC Name 4-chloronaphthalen-1-ol
SMILES C1=CC=C2C(=C1)C(=CC=C2Cl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 117°C to 120°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC10H6OH
Quantity 100 g
Beilstein 06, 611
Solubility Information Solubility in water: negligible in water. Other solubilities: soluble in methanol and 8% 2,2′oxydiethanol
Formula Weight 178.62
Percent Purity 97%
Physical Form Needles or Powder
Chemical Name or Material 4-Chloro-1-naphthol, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.