missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde, 95+%, Thermo Scientific™

Product Code. 10089321
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089321 250 mg Amber Glass Bottle 250mg
10581322 1 g Amber Glass Bottle 1g
10245614 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10089321 Supplier Thermo Scientific Maybridge Supplier No. RF03946CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-81-6
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00103234
InChI Key MLGDASSMRFJUMZ-UHFFFAOYSA-N
Synonym 4-chloro-1-methyl-1h-pyrazole-3-carbaldehyde, 4-chloro-1-methyl-1h-pyrazole-3-carboxaldehyde, 4-chloro-1-methylpyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde, 4-chloro-1-methyl-9ci
PubChem CID 2782153
SMILES CN1C=C(Cl)C(C=O)=N1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Formula Weight 144.56
Percent Purity 95%
Chemical Name or Material 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.