missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butanol, 85%, balance THF and HCl

Product Code. 10499232 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10499232 10 g Glass bottle 10g
10634082 100 g Glass bottle 100g
10118033 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10499232 Supplier Thermo Scientific Acros Supplier No. 356310100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-51-8 | C4H9ClO | 108.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-51-8
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00002967
InChI Key HXHGULXINZUGJX-UHFFFAOYSA-N
Synonym 4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
PubChem CID 13569
ChEBI CHEBI:87310
IUPAC Name 4-chlorobutan-1-ol
SMILES OCCCCCl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0890g/mL
Boiling Point 58.0°C to 60.0°C (3.0 mmHg)
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)4OH
Refractive Index 1.4520 to 1.4560
Quantity 10 g
Beilstein 01, II, 398
Specific Gravity 1.089
Formula Weight 108.57
Percent Purity 85%
Physical Form Liquid
Chemical Name or Material 4-Chloro-1-butanol, Balance THF and HCl
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-1-butanol, balance THF and HCl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.