missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,8-naphthalic anhydride, 94%

Product Code. p-7044254
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11377465

Brand: Thermo Scientific Alfa Aesar L05508.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4053-08-1 | C12H5ClO3 | 232.62 g/mol

4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4053-08-1
Molecular Formula C12H5ClO3
Molecular Weight (g/mol) 232.62
MDL Number MFCD00006928
InChI Key UJEUBSWHCGDJQU-UHFFFAOYSA-N

Specifications

Melting Point 208°C to 214°C
Quantity 25 g
Beilstein 178072
Sensitivity Moisture sensitive
Synonym 4-chloro-1,8-naphthalic anhydride, 4-chloronaphthalic anhydride, 6-chlorobenzo de isochromene-1,3-dione, 4-chloronapthalic anhydride, 6-chloro-1h,3h-benzo de isochromene-1,3-dione, 4-chloronaphthalene-1,8-dicarboxylic anhydride, 1h,3h-naphtho 1,8-cd pyran-1,3-dione, 6-chloro, naphthalic anhydride, 4-chloro-7ci,8ci, 6-chloro-1h,3h-naphtho 1,8-cd pyran-1,3-dione
Solubility Information Hydrolyzes in water.
SMILES C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Cl
PubChem CID 77672
Formula Weight 232.63
Percent Purity 94%
Chemical Name or Material 4-Chloro-1,8-naphthalic anhydride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.