missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10651483
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10651483 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10651483 Supplier Thermo Scientific Maybridge Supplier No. KM02255DA
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175201-94-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H8ClN3O2
MDL Number MFCD00067959
Quantity 1 g
Synonym 4-chloro-1,3-dimethyl-1h-pyrazolo 3,4-b pyridine-5-carboxylic acid, 4-chloro-1,3-dimethylpyrazolo 3,4-b pyridine-5-carboxylic acid, 4-chloro-1,3-dimethyl-1h-pyrazol 3,4-b pyridine-5-carboxylic acid, 1h-pyrazolo 3,4-b pyridine-5-carboxylicacid, 4-chloro-1,3-dimethyl, 4-chloro-1,3-dimethylpyrazolo 3,4-b pyridine, 4-chloro-1,3-dimethylpyrazolo 5,4-b pyridine-5-carboxylic acid, buttpark 30\08-34, 4-chloro-1,3-dimethyl-1h-pyrazolo 3,4-b pyridine-5, 4-chloro-1,3-dimethylpyraazolo 3,4-b pyridine-5-carboxylic acid
InChI Key LHEUUYZQYKWDSY-UHFFFAOYSA-N
SMILES CN1N=C(C)C2=C(Cl)C(=CN=C12)C(O)=O
IUPAC Name 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylic acid
Molecular Weight (g/mol) 225.63
PubChem CID 614513
Formula Weight 225.63
Percent Purity 97%
Chemical Name or Material 4-chloro-1, 3-dimethyl-1H-pyrazolo[3, 4-b]pyridine-5-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.