missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro(1,1'-biphenyl)-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10209502
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10209502

Brand: Thermo Scientific Maybridge MO07005EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20443-74-7
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.154
MDL Number MFCD01631918
InChI Key NWYUSJMIHFIMTA-UHFFFAOYSA-N
Synonym 4'-chlorobiphenyl-4-sulfonyl chloride, 4-4-chlorophenyl benzenesulfonyl chloride, 4'-chloro-biphenyl-4-sulfonyl chloride, 4'-chloro-1,1'-biphenyl-4-sulfonyl chloride, 4'-chloro-4-biphenylylsulphonyl chloride, 4'-chloro 1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-chloro, 4'-chlorobiphenyl4-sulfonyl chloride, 4-chlorobiphenyl-4'-sulfonyl chloride, 4'-chloro biphenyl-4-sulfonyl chloride
PubChem CID 2794745
IUPAC Name 4-(4-chlorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 287.17
Percent Purity 97%
Chemical Name or Material 4';-Chloro[1, 1';-biphenyl]-4-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.