missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxyphenylboronic acid, 97%

Product Code. 10556261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10556261 1 g Glass bottle 1g
10566261 5 g Glass bottle 5g
10626194 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10556261 Supplier Thermo Scientific Acros Supplier No. 344400010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14047-29-1 | C7H7BO4 | 165.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14047-29-1
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.94
MDL Number MFCD00151801
InChI Key SIAVMDKGVRXFAX-UHFFFAOYSA-N
Synonym 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid, p-carboxyphenylboronic acid, 4-phenylester boronic acid, 4-carboxyphenyl boronic acid, chembl82324
PubChem CID 312183
IUPAC Name 4-boronobenzoic acid
SMILES OB(O)C1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 238.0°C to 240.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOOCC6H4B(OH)2
Quantity 1 g
Solubility Information Solubility in water: 25g/L.
Formula Weight 165.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Carboxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Carboxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.