missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11396959
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396959 5 g 5g
11386959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396959 Supplier Thermo Scientific Alfa Aesar Supplier No. H26825.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 180516-87-4 | Molecular Formula: C13H17BO4 | Molecular Weight: 248.09 g/mol

4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzeneboronic acid pinacol ester is used to improvement of transition metal-mediated carbon-carbon as well as carbon-heteroatom cross-coupling reactions as a labeling feature with the focus on the preparation of radio labeled compounds for molecular imaging.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180516-87-4
Molecular Formula C13H17BO4
Molecular Weight (g/mol) 248.09
MDL Number MFCD01863710
InChI Key IYDKBQIEOBXLTP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid pinacol ester, 4-carboxylphenylboronic acid pinacol ester, 4-carboxyphenylboronic acid, pinacol ester, 4-carboxyphenylboronicacidpinacolester, 4-carboxybenzeneboronic acid pinacol ester, 4-carboxyphenylboronic acid pinacolate, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxyphenylboronic acid,pinacol ester, 4-carboxyphenyl boronic acid, pinacol ester
PubChem CID 2734621
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 227°C to 231°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 248.08
Percent Purity 97%
Chemical Name or Material 4-Carboxybenzeneboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-Carboxybenzeneboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Carboxylphenylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.