missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzaldehyde, 98%

Product Code. p-7022473
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11462338

Brand: Thermo Scientific Alfa Aesar A15277.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-66-9 | C8H6O3 | 150.133 g/mol

4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

Solubility
Soluble in water, methanol, DMSO, ether, and chloroform.

Notes
Store away from air and strong bases. Air Sensitive. Incompatible with bases and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-66-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00006951
InChI Key GOUHYARYYWKXHS-UHFFFAOYSA-N
Synonym 4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
PubChem CID 12088
IUPAC Name 4-formylbenzoic acid
SMILES C1=CC(=CC=C1C=O)C(=O)O

Specifications

Melting Point ∼245°C
Odor Odorless
Quantity 10 g
Beilstein 471734
Sensitivity Air Sensitive
Solubility Information Soluble in water,methanol,DMSO,ether,and chloroform.
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material 4-Carboxybenzaldehyde
compliance-icons compliance-icons
Product Identifier
  • 4-Carboxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4-formyl-benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.