missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzaldehyde, 98%

Product Code. 11482338
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482338 250 g 250g
11462338 10 g 10g
11472338 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482338 Supplier Thermo Scientific Alfa Aesar Supplier No. A15277.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 619-66-9 | C8H6O3 | 150.133 g/mol

4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

Solubility
Soluble in water, methanol, DMSO, ether, and chloroform.

Notes
Store away from air and strong bases. Air Sensitive. Incompatible with bases and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-66-9
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00006951
InChI Key GOUHYARYYWKXHS-UHFFFAOYSA-N
Synonym 4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
PubChem CID 12088
IUPAC Name 4-formylbenzoic acid
SMILES C1=CC(=CC=C1C=O)C(=O)O

Specifications

Melting Point ∼245°C
Odor Odorless
Quantity 250 g
Beilstein 471734
Sensitivity Air Sensitive
Solubility Information Soluble in water,methanol,DMSO,ether,and chloroform.
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material 4-Carboxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.