missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-3-fluorobenzeneboronic acid, 98%

Product Code. 15430038
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430038 1 g 1g
15440038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430038 Supplier Thermo Scientific Alfa Aesar Supplier No. H53285.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120153-08-4 | C7H6BFO4 | 183.929 g/mol

4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120153-08-4
Molecular Formula C7H6BFO4
Molecular Weight (g/mol) 183.929
MDL Number MFCD01114671
InChI Key CZDWJVSOQOMYGC-UHFFFAOYSA-N
Synonym 4-carboxy-3-fluorophenylboronic acid, 4-carboxy-3-fluorobenzeneboronic acid, 4-dihydroxyboranyl-2-fluorobenzoic acid, 4-carboxy-3-fluorophenyl boronic acid, benzoic acid, 4-borono-2-fluoro, 3-fluoro-4-carboxyphenylboronic acid, pubchem5148, acmc-209a6f, benzoic acid,4-borono-2-fluoro, 3-fluoro-4-carboxy-phenylboronic acid
PubChem CID 2763201
IUPAC Name 4-borono-2-fluorobenzoic acid
SMILES B(C1=CC(=C(C=C1)C(=O)O)F)(O)O

Specifications

Melting Point 236°C to 240°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 183.93
Percent Purity 98%
Chemical Name or Material 4-Carboxy-3-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.