missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15406946
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1050423-87-4
Molecular Formula C13H16BFO4
Molecular Weight (g/mol) 266.075
MDL Number MFCD12756466
InChI Key JSQNZNXVVRPGTD-UHFFFAOYSA-N
Synonym 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxy-2-fluorophenylboronic acid pinacol ester, 4-carboxy-2-fluorobenzeneboronic acid pinacol ester, 4-carboxy-2-fluorophenylboronic acid, pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxy-2-fluorophenylboronicacidpinacolester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoicacid, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoic acid
PubChem CID 46739808
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)O)F

Specifications

Melting Point 199°C to 204°C
Quantity 5 g
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'