missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15412238
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
851135-07-4
C7H6BFO4
183.929
MFCD08458473
TVNVDRMGOOLVOX-UHFFFAOYSA-N
4-carboxy-2-fluorophenylboronic acid, 4-carboxy-2-fluorobenzeneboronic acid, 4-dihydroxyboranyl-3-fluorobenzoic acid, benzoic acid, 4-borono-3-fluoro, 4-carboxy-2-fluorophenylboronicacid, acmc-209q4r, 4-borono-3-fluoro-benzoic acid, 4-?borono-?3-?fluoro-benzoic acid, 4-carboxy-2-fluorobezeneboronic acid, 3-fluoro-4-dihydroxyboryl benzoic acid
23134177
4-borono-3-fluorobenzoic acid
B(C1=C(C=C(C=C1)C(=O)O)F)(O)O
This item is not returnable. View return policy
851135-07-4
C7H6BFO4
183.929
MFCD08458473
TVNVDRMGOOLVOX-UHFFFAOYSA-N
4-carboxy-2-fluorophenylboronic acid, 4-carboxy-2-fluorobenzeneboronic acid, 4-dihydroxyboranyl-3-fluorobenzoic acid, benzoic acid, 4-borono-3-fluoro, 4-carboxy-2-fluorophenylboronicacid, acmc-209q4r, 4-borono-3-fluoro-benzoic acid, 4-?borono-?3-?fluoro-benzoic acid, 4-carboxy-2-fluorobezeneboronic acid, 3-fluoro-4-dihydroxyboryl benzoic acid
23134177
4-borono-3-fluorobenzoic acid
B(C1=C(C=C(C=C1)C(=O)O)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 183.93
Percent Purity 97%
Chemical Name or Material 4-Carboxy-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'