missing translation for 'onlineSavingsMsg'
Learn More

4-Carbethoxyimidazole, TRC

Product Code. 30359050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359050 1 g -
30362995 25 g 25g
30363045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359050 Supplier Toronto Research Chemicals Supplier No. TRCC1761101G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23785-21-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
Synonym 1H-Imidazole-5-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)imidazole, C 751, C 751 (pharmaceutical), Ethyl 1H-Imidazole-4-carboxylate, Ethyl 4-imidazolecarboxylate, NSC 191283
IUPAC Name ethyl 1H-imidazole-4-carboxylate
SMILES CCOC(=O)c1c[nH]cn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H8N2O2/c1-2-10-6(9)5-3-7-4-8-5/h3-4H,2H2,1H3,(H,7,8)
Formula Weight 140.0586
Chemical Name or Material 4-Carbethoxyimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.