missing translation for 'onlineSavingsMsg'
Learn More

4-Butylaniline, 97%

Product Code. 10261410 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10261410

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-13-2 | C10H15N | 149.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-13-2
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.23
InChI Key OGIQUQKNJJTLSZ-UHFFFAOYSA-N
Synonym 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine
PubChem CID 7694
IUPAC Name 4-butylaniline
SMILES CCCCC1=CC=C(C=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.9450g/mL
Boiling Point 133.0°C to 134.0°C (14.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3C6H4NH2
Refractive Index 1.535
Quantity 25 g
Beilstein 12,I,503
Specific Gravity 0.945
Formula Weight 149.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Butylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Butylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.